Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hnrnpc
Entrez Gene ID
406352
Gene Name
heterogeneous nuclear ribonucleoprotein C
Gene Aliases
bZ1G18.4, fb57h12, fi16b11, wu:fb57h12, wu:fi16b11, zgc:55701
Chromosome Location
Chr.2: 68126 - 77059 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213079.1 NP_998244.1 5 - 6 601 83
XM_021479568.1 XP_021335243.1 5 - 6 692 83
XM_021479569.1 XP_021335244.1 6 - 7 688 83
GenBank mRNA BC044458.1 - 5 - 6 601 83
BC097078.1 - 5 - 6 686 83

View Full Product Details