Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmbim4
Entrez Gene ID
406412
Gene Name
transmembrane BAX inhibitor motif containing 4
Gene Aliases
wu:fb34a09, zgc:64112
Chromosome Location
Chr.4: 12971250 - 12978931 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213138.2 NP_998303.2 5 - 6 595 78
GenBank mRNA BC057432.1 - 5 - 6 535 78
EH442390.1 - 5 - 6 602 78
EH490666.1 - 5 - 6 597 78

View Full Product Details