Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tax1bp3
Entrez Gene ID
406779
Gene Name
Tax1 (human T-cell leukemia virus type I) binding protein 3
Gene Aliases
wu:fb37e03, wu:fc23g08, zgc:55513
Chromosome Location
Chr.11: 6136260 - 6145158 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213470.1 NP_998635.1 2 - 3 272 78
GenBank mRNA BC042322.1 - 2 - 3 272 78

View Full Product Details