Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sult6b1
Entrez Gene ID
322462
Gene Name
sulfotransferase family, cytosolic, 6b, member 1
Gene Aliases
sb:cb89, wu:fb62a12, zgc:111843
Chromosome Location
Chr.11: 43375428 - 43387747 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214686.2 NP_999851.2 5 - 6 714 70
GenBank mRNA AY180110.2 - 5 - 6 644 70
BC093112.1 - - 700 70

View Full Product Details