Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT2
Entrez Gene ID
100056092
Gene Name
Wnt family member 2
Gene Aliases
-
Chromosome Location
Chr.4: 74570722 - 74613286 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114150.1 NP_001107622.1 1 - 2 387 78

View Full Product Details