Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC27
Entrez Gene ID
100055919
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ
Chromosome Location
Chr.15: 71918808 - 71951040 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195523.1 NP_001182452.1 1 - 2 80 139

View Full Product Details