Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDN2
Entrez Gene ID
100033847
Gene Name
endothelin 2
Gene Aliases
-
Chromosome Location
Chr.2: 16841843 - 16847507 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081823.2 NP_001075292.1 3 - 4 443 81
GenBank mRNA AB079136.1 - 3 - 4 444 81

View Full Product Details