Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSN3
Entrez Gene ID
100033983
Gene Name
casein kappa
Gene Aliases
-
Chromosome Location
Chr.3: 66469756 - 66480050 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081884.1 NP_001075353.1 1 - 2 76 125
GenBank mRNA AY040863.1 - 1 - 2 76 125

View Full Product Details