Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY96
Entrez Gene ID
100034040
Gene Name
lymphocyte antigen 96
Gene Aliases
-
Chromosome Location
Chr.9: 12908086 - 12934704 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081898.1 NP_001075367.1 4 - 5 422 92
GenBank mRNA AY398685.1 - 4 - 5 422 92

View Full Product Details