Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPMT
Entrez Gene ID
100034066
Gene Name
thiopurine S-methyltransferase
Gene Aliases
-
Chromosome Location
Chr.20: 17518382 - 17538083 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081905.1 NP_001075374.1 6 - 7 583 133
XM_023624007.1 XP_023479775.1 - 811 133
XM_023624008.1 XP_023479776.1 - 815 133
GenBank mRNA AY827076.1 - 6 - 7 583 133

View Full Product Details