Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GM2A
Entrez Gene ID
100034082
Gene Name
GM2 ganglioside activator
Gene Aliases
-
Chromosome Location
Chr.14: 26184319 - 26197304 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081912.1 NP_001075381.1 1 - 2 91 66
GenBank mRNA DQ167467.1 - 1 - 2 91 66

View Full Product Details