Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECATH-3
Entrez Gene ID
100034135
Gene Name
myeloid cathelicidin 3
Gene Aliases
eCATH-1
Chromosome Location
Chr.16: 40267791 - 40269970 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081930.2 NP_001075399.1 3 - 4 385 127
GenBank mRNA AJ224929.1 - 3 - 4 385 127

View Full Product Details