Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGF1
Entrez Gene ID
100034198
Gene Name
insulin like growth factor 1
Gene Aliases
-
Chromosome Location
Chr.28: 27225722 - 27301668 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082498.2 NP_001075967.2 3 - 4 417 63
XM_005606472.3 XP_005606529.1 - 1011 63
GenBank mRNA EU018459.1 - 3 - 4 417 63

View Full Product Details