Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL1A
Entrez Gene ID
100064969
Gene Name
interleukin 1 alpha
Gene Aliases
-
Chromosome Location
Chr.15: 16360699 - 16369812 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082500.2 NP_001075969.2 2 - 3 103 112
GenBank mRNA CD466534.1 - 2 - 3 103 112
D42146.1 - 1 - 2 55 112
U92480.1 - 2 - 3 82 112

View Full Product Details