Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ILT11A
Entrez Gene ID
100036551
Gene Name
immunoglobulin-like transcript 11 A
Gene Aliases
-
Chromosome Location
Chr.10: 23728087 - 23857862 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083593.1 NP_001077062.1 2 - 3 173 65
XM_014735367.2 XP_014590853.1 - 379 65
XM_014736781.2 XP_014592267.1 - 380 65
XM_023649350.1 XP_023505118.1 - 552 65
XM_023649351.1 XP_023505119.1 - 627 65
XM_023649352.1 XP_023505120.1 - 762 65
XM_023649354.1 XP_023505122.1 - 687 65
XM_023649355.1 XP_023505123.1 - 550 65
XM_023649356.1 XP_023505124.1 - 451 65
XM_023649357.1 XP_023505125.1 - 527 65
XM_023649360.1 XP_023505128.1 - 558 65
XM_023649361.1 XP_023505129.1 - 558 65
XM_023649363.1 XP_023505131.1 - 558 65
XM_023649364.1 XP_023505132.1 - 555 65
XM_023649365.1 XP_023505133.1 - 383 65
XM_023649366.1 XP_023505134.1 - 382 65
GenBank mRNA AB120403.1 - 2 - 3 173 65
AB120404.1 - 2 - 3 173 65
AB120406.1 - 1 - 2 91 65
AB120407.1 - 1 - 2 89 65

View Full Product Details