Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MILL
Entrez Gene ID
100034249
Gene Name
MHC I like leukocyte pseudogene
Gene Aliases
-
Chromosome Location
Chr.10: 16713512 - 16723177 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_036560.1 3 - 4 630 120
GenBank mRNA AB126064.1 - 3 - 4 630 120

View Full Product Details