Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKDC
Entrez Gene ID
791233
Gene Name
protein kinase, DNA-activated, catalytic subunit
Gene Aliases
DNA-PK, DNAPK
Chromosome Location
Chr.9: 36236670 - 36438603 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163858.2 NP_001157330.1 30 - 31 3604 153
XM_023648324.1 XP_023504092.1 - 3672 153
XM_023648325.1 XP_023504093.1 - 3782 153
XM_023648326.1 XP_023504094.1 - 3519 153
XM_023648328.1 XP_023504096.1 - 3672 153
GenBank mRNA AF448228.1 - 30 - 31 3524 153

View Full Product Details