Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRKDC
Entrez Gene ID
791233
Gene Name
protein kinase, DNA-activated, catalytic subunit
Gene Aliases
DNA-PK, DNAPK
Chromosome Location
Chr.9: 36236670 - 36438603 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163858.2 NP_001157330.1 75 - 76 10781 130
XM_023648324.1 XP_023504092.1 - 10849 130
XM_023648325.1 XP_023504093.1 - 10959 130
XM_023648326.1 XP_023504094.1 - 10696 130
GenBank mRNA AF448228.1 - 75 - 76 10698 130

View Full Product Details