Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNS
Entrez Gene ID
100034131
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Gene Aliases
G6S
Chromosome Location
Chr.6: 81638600 - 81675726 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001492151.6 XP_001492201.6 - 1154 87
XM_023643942.1 XP_023499710.1 - 1154 87
GenBank mRNA AF510336.1 - 8 - 9 1042 87

View Full Product Details