Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GFAP
Entrez Gene ID
100033970
Gene Name
glial fibrillary acidic protein
Gene Aliases
-
Chromosome Location
Chr.11: 18815101 - 18824260 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163861.1 NP_001157333.1 3 - 4 618 126
XM_005597018.3 XP_005597075.1 - 636 126
XM_023651587.1 XP_023507355.1 - 636 126
GenBank mRNA AJ551396.1 - 2 - 3 151 126

View Full Product Details