Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STS
Entrez Gene ID
100034102
Gene Name
steroid sulfatase
Gene Aliases
-
Chromosome Location
Chr.X: 4626346 - 4805915 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163953.1 NP_001157425.1 8 - 9 1364 116
XM_014728733.2 XP_014584219.1 - 2339 116
XM_023633178.1 XP_023488946.1 - 1698 116
GenBank mRNA DQ367333.1 - 5 - 6 610 116

View Full Product Details