Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNFSF12
Entrez Gene ID
100061872
Gene Name
TNF superfamily member 12
Gene Aliases
TNLG4A
Chromosome Location
Chr.11: 50865106 - 50873355 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205160.1 NP_001192089.1 4 - 5 435 100
XM_005597024.3 XP_005597081.1 - 455 100

View Full Product Details