Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM34
Entrez Gene ID
100568304
Gene Name
tripartite motif containing 34
Gene Aliases
-
Chromosome Location
Chr.7: 76637162 - 76652166 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205193.1 NP_001192122.1 5 - 6 870 80
XM_005611916.3 XP_005611973.1 - 1159 80
XM_005611917.3 XP_005611974.1 - 1308 80

View Full Product Details