Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GTF2A1L
Entrez Gene ID
100534009
Gene Name
general transcription factor IIA subunit 1 like
Gene Aliases
-
Chromosome Location
Chr.15: 51639377 - 51683910 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204845.1 NP_001191774.1 2 - 3 164 82
XM_023618425.1 XP_023474193.1 - 223 82

View Full Product Details