Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TOMM20
Entrez Gene ID
426309
Gene Name
translocase of outer mitochondrial membrane 20
Gene Aliases
TOMM20L
Chromosome Location
Chr.3: 38288127 - 38300528 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_423972.6 XP_423972.3 1 - 2 221 98
GenBank mRNA BX929316.1 - 1 - 2 208 98
BX930714.2 - 1 - 2 216 98
BX930835.2 - 1 - 2 225 98
BX930873.1 - 1 - 2 201 98
BX932356.1 - 1 - 2 198 98

View Full Product Details