Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDN2
Entrez Gene ID
419559
Gene Name
endothelin 2
Gene Aliases
endothelin-2
Chromosome Location
Chr.23: 1063330 - 1068649 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198655.3 NP_001185584.1 2 - 3 334 89
GenBank mRNA BX929792.1 - 2 - 3 227 89
BX932521.1 - 2 - 3 335 89
BX934036.2 - 1 - 2 93 89

View Full Product Details