Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDKN3
Entrez Gene ID
423562
Gene Name
cyclin dependent kinase inhibitor 3
Gene Aliases
-
Chromosome Location
Chr.5: 56665665 - 56671678 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252162.1 NP_001239091.1 4 - 5 521 82
GenBank mRNA BX929389.2 - 1 - 2 653 82
BX929945.1 - 4 - 5 315 82
BX933305.1 - 3 - 4 440 82
CR353075.1 - 3 - 4 390 82

View Full Product Details