Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL3
Entrez Gene ID
423865
Gene Name
ADP ribosylation factor like GTPase 3
Gene Aliases
-
Chromosome Location
Chr.6: 24255064 - 24283167 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025151664.1 XP_025007432.1 2 - 3 321 116
XM_421730.6 XP_421730.1 2 - 3 215 116
GenBank mRNA BX929388.1 - 1 - 2 167 116
BX930620.1 - 2 - 3 209 116

View Full Product Details