Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP1LC3C
Entrez Gene ID
421504
Gene Name
microtubule associated protein 1 light chain 3 gamma
Gene Aliases
-
Chromosome Location
Chr.3: 35797595 - 35799581 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004935459.3 XP_004935516.1 2 - 3 390 70
XM_419549.6 XP_419549.2 3 - 4 454 70
GenBank mRNA BX930693.2 - 3 - 4 221 70
BX930772.1 - 2 - 3 289 70
BX932218.1 - 1 - 2 413 70
BX932346.1 - 3 - 4 459 70
BX933244.1 - 2 - 3 293 70
BX934613.1 - 2 - 3 295 70

View Full Product Details