Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDDC1
Entrez Gene ID
423020
Gene Name
Parkinson disease 7 domain containing 1
Gene Aliases
GATD1
Chromosome Location
Chr.5: 691717 - 705919 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199565.1 NP_001186494.1 6 - 7 582 83
NM_001199566.1 NP_001186495.1 6 - 7 582 83
XM_015286220.2 XP_015141706.2 6 - 7 583 83
XM_025150660.1 XP_025006428.1 6 - 7 1103 83
XM_025150661.1 XP_025006429.1 6 - 7 1102 83
XM_025150662.1 XP_025006430.1 6 - 7 1101 83
XM_025150663.1 XP_025006431.1 6 - 7 973 83
XM_025150664.1 XP_025006432.1 6 - 7 532 83
GenBank mRNA AM065422.1 - 6 - 7 582 83
BU253847.1 - 4 - 5 330 83
BX929358.2 - 6 - 7 575 83
BX932402.1 - 6 - 7 577 83
BX932999.2 - 3 - 4 219 83
BX934566.2 - 5 - 6 596 83

View Full Product Details