Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS21
Entrez Gene ID
419228
Gene Name
ribosomal protein S21
Gene Aliases
-
Chromosome Location
Chr.20: 7935113 - 7938522 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_417405.6 XP_417405.4 5 - 6 309 80
GenBank mRNA BX932886.1 - 4 - 5 277 80

View Full Product Details