Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM189A2
Entrez Gene ID
427245
Gene Name
family with sequence similarity 189 member A2
Gene Aliases
C9orf61
Chromosome Location
Chr.Z: 34684448 - 34709655 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025144867.1 XP_025000635.1 5 - 6 546 122
XM_025144868.1 XP_025000636.1 5 - 6 498 122
XM_424828.6 XP_424828.4 5 - 6 814 122
GenBank mRNA BX933855.1 - 3 - 4 287 122

View Full Product Details