Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN13
Entrez Gene ID
420595
Gene Name
tetraspanin 13
Gene Aliases
TM4SF13, tetraspanin-13
Chromosome Location
Chr.2: 28543307 - 28560140 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199612.1 NP_001186541.1 5 - 6 689 75
XM_015281410.2 XP_015136896.1 6 - 7 739 75
GenBank mRNA BX933986.1 - 5 - 6 563 75
BX934713.2 - 5 - 6 635 75
CN210835.1 - 5 - 6 689 75

View Full Product Details