Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM32A
Entrez Gene ID
420150
Gene Name
family with sequence similarity 32 member A
Gene Aliases
-
Chromosome Location
Chr.28: 4518104 - 4519229 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_418266.5 XP_418266.2 2 - 3 239 65
GenBank mRNA BX934951.1 - 2 - 3 239 65
CR353360.1 - 2 - 3 228 65

View Full Product Details