Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATSL2
Entrez Gene ID
417483
Gene Name
GATS protein like 2
Gene Aliases
CASTOR2, GATS, GATSL1
Chromosome Location
Chr.19: 2690346 - 2784509 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281807.1 NP_001268736.1 3 - 4 691 74
XM_015295702.2 XP_015151188.1 4 - 5 924 74
GenBank mRNA BX950387.1 - 3 - 4 441 74
BX950514.1 - 3 - 4 375 74
CR405722.1 - 3 - 4 485 74

View Full Product Details