Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MICU3
Entrez Gene ID
422736
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
Chromosome Location
Chr.4: 63212802 - 63266757 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015285646.2 XP_015141132.1 3 - 4 766 153
GenBank mRNA CR338835.1 - 1 - 2 309 153

View Full Product Details