Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DBX2
Entrez Gene ID
417801
Gene Name
developing brain homeobox 2
Gene Aliases
CHoxE
Chromosome Location
Chr.1: 30657836 - 30682019 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276354.1 NP_001263283.1 1 - 2 398 95
XM_025147738.1 XP_025003506.1 1 - 2 805 95
GenBank mRNA BX929917.2 - 1 - 2 157 95

View Full Product Details