Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CYP2D6
- Entrez Gene ID
-
417981
- Gene Name
- cytochrome P450 family 2 subfamily D member 6
- Gene Aliases
-
CYP2D49, CYP2D6c
- Chromosome Location
-
Chr.1: 49408430 - 49417609 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX930713.2 |
- |
2 - 3 |
506 |
150 |
View Full Product Details