The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_415925.5 | XP_415925.1 | 1 - 2 | 210 | 139 |
GenBank mRNA | BX929616.2 | - | 1 - 2 | 153 | 139 |
BX932710.2 | - | 1 - 2 | 424 | 139 |