Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC7
Entrez Gene ID
420704
Gene Name
exosome component 7
Gene Aliases
-
Chromosome Location
Chr.2: 43050870 - 43070357 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252154.1 NP_001239083.1 2 - 3 184 68
GenBank mRNA BU426698.1 - 2 - 3 187 68
BX932890.2 - 2 - 3 184 68
BX933850.1 - 2 - 3 159 68
BX935756.1 - 2 - 3 182 68

View Full Product Details