Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC7
Entrez Gene ID
420704
Gene Name
exosome component 7
Gene Aliases
-
Chromosome Location
Chr.2: 43050870 - 43070357 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252154.1 NP_001239083.1 4 - 5 442 73
GenBank mRNA BX932890.2 - 4 - 5 442 73
BX933850.1 - 4 - 5 417 73
BX935756.1 - 4 - 5 440 73

View Full Product Details