Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM65
Entrez Gene ID
417367
Gene Name
tripartite motif containing 65
Gene Aliases
-
Chromosome Location
Chr.18: 4625152 - 4629907 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_415625.5 XP_415625.2 5 - 6 2515 54
GenBank mRNA BX934912.2 - 1 - 2 27 54

View Full Product Details