Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- COA7
- Entrez Gene ID
-
429105
- Gene Name
- cytochrome c oxidase assembly factor 7 (putative)
- Gene Aliases
-
C1orf163, C8H1orf163, SELRC1
- Chromosome Location
-
Chr.8: 24856006 - 24858084 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BX935415.2 |
- |
1 - 2 |
123 |
77 |
View Full Product Details