Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COQ7
Entrez Gene ID
416609
Gene Name
coenzyme Q7, hydroxylase
Gene Aliases
-
Chromosome Location
Chr.14: 8869571 - 8882281 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277172.1 NP_001264101.1 1 - 2 96 56
XM_015294222.2 XP_015149708.1 1 - 2 160 56
XM_015294223.2 XP_015149709.1 1 - 2 160 56
XM_025155105.1 XP_025010873.1 1 - 2 160 56
XM_025155106.1 XP_025010874.1 1 - 2 160 56
GenBank mRNA BX933254.1 - 1 - 2 94 56
BX935091.1 - 1 - 2 91 56
BX935416.1 - 1 - 2 101 56
CR385437.1 - 1 - 2 96 56

View Full Product Details