Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEPTIN8
Entrez Gene ID
416333
Gene Name
septin 8
Gene Aliases
SEPT8, Sep-08
Chromosome Location
Chr.13: 17690033 - 17723569 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX936065.1 - 1 - 2 53 69
BX950469.2 - 3 - 4 409 69
CR405735.1 - 2 - 3 287 69

View Full Product Details