Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCUN1D2
Entrez Gene ID
418740
Gene Name
defective in cullin neddylation 1 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 138728030 - 138749323 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015277897.2 XP_015133383.1 5 - 6 1297 162
XM_015277898.2 XP_015133384.1 5 - 6 685 162
XM_015277899.2 XP_015133385.1 6 - 7 823 162
XM_025146715.1 XP_025002483.1 5 - 6 871 162
GenBank mRNA BX932972.1 - 3 - 4 366 162
CR386062.1 - 2 - 3 222 162
CR523518.1 - 5 - 6 637 162

View Full Product Details