Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHLDA2
Entrez Gene ID
423088
Gene Name
pleckstrin homology like domain family A member 2
Gene Aliases
-
Chromosome Location
Chr.5: 13345377 - 13346840 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199595.1 NP_001186524.1 1 - 2 514 122
GenBank mRNA BX929348.1 - 1 - 2 458 122
BX950502.2 - 1 - 2 603 122

View Full Product Details