Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WBP1L
Entrez Gene ID
423867
Gene Name
WW domain binding protein 1-like
Gene Aliases
C10orf26, C6H10ORF26, OPA1L, OPAL1
Chromosome Location
Chr.6: 24289588 - 24339659 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004942244.3 XP_004942301.1 4 - 4 1562 71
XM_015288736.2 XP_015144222.1 3 - 3 1655 71
XM_015288740.1 XP_015144226.1 4 - 4 1458 71
XM_015288742.2 XP_015144228.1 4 - 4 1711 71
XM_025151875.1 XP_025007643.1 6 - 6 1949 71
XM_025151876.1 XP_025007644.1 5 - 5 1853 71
XM_025151877.1 XP_025007645.1 5 - 5 1681 71
XM_025151878.1 XP_025007646.1 5 - 5 1643 71
XM_421732.6 XP_421732.1 4 - 4 1664 71
GenBank mRNA BX931788.2 - 1 - 1 756 71

View Full Product Details