Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIH1D3
Entrez Gene ID
422184
Gene Name
PIH1 domain containing 3
Gene Aliases
C4HXORF41, CXorf41, NYSAR97
Chromosome Location
Chr.4: 1905206 - 1913527 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_420180.6 XP_420180.3 6 - 7 596 100
GenBank mRNA BX934346.2 - 6 - 7 580 100

View Full Product Details