Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UGT1A1
Entrez Gene ID
424028
Gene Name
UDP glucuronosyltransferase family 1 member A1
Gene Aliases
UGT1A10, UGT1A6, UGT1A7, UGT1A8, UGT1A9
Chromosome Location
Chr.7: 5831606 - 5902469 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015289254.2 XP_015144740.2 3 - 4 2050 102
XM_025152657.1 XP_025008425.1 2 - 3 1974 102
GenBank mRNA BX936208.2 - 1 - 2 512 102

View Full Product Details